Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011592.2 | NP_035722.2 | 7 | 764 | ||
GenBank mRNA | AK153866.1 | - | 7 | 769 | ||
BC023454.1 | - | 7 | 740 | |||
BC043479.1 | - | 7 | 744 | |||
BC110677.1 | - | 7 | 762 | 4979986 | ||
BC117523.1 | - | 7 | 754 | 40105563 | ||
BC117524.1 | - | 7 | 754 | 40105561 | ||
U69898.1 | - | 7 | 754 |