Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011602.5 | NP_035732.2 | 49 | 7035 | ||
GenBank mRNA | AK122420.2 | - | 49 | 6661 | ||
AK147228.1 | - | 49 | 6606 | |||
AK170630.1 | - | 15 | 2078 | |||
AK171322.1 | - | 15 | 2078 | |||
BC018557.1 | - | 13 | 1743 | |||
BC121787.1 | - | 19 | 2547 | |||
BC150810.1 | - | 49 | 6706 | 9087721 | ||
X56123.1 | - | 49 | 6707 |