Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011648.5 | NP_035778.3 | 10 | 1035 | ||
GenBank mRNA | AK029084.1 | - | 10 | 1001 | ||
BC086691.1 | - | 10 | 964 | 30682686 | ||
BC092523.1 | - | 10 | 977 | 30650002 | ||
U02601.1 | - | 10 | 1017 | |||
U02602.1 | - | 10 | 1017 |