Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011695.2 | NP_035825.1 | 3, 4 | 237 | ||
GenBank mRNA | AK012359.1 | - | 3, 4 | 235 | ||
AK078555.1 | - | 3, 4 | 241 | |||
AK150504.1 | - | 3, 4 | 263 | |||
AK150627.1 | - | 3, 4 | 263 | |||
AK150940.1 | - | 3, 4 | 255 | |||
AK152274.1 | - | 3, 4 | 235 | |||
AK159561.1 | - | 3, 4 | 472 | |||
AK168199.1 | - | 3, 4 | 208 | |||
AK168328.1 | - | 3, 4 | 234 | |||
AK191978.1 | - | 1, 2 | 86 | |||
BC003731.1 | - | 3, 4 | 237 | 3601668 | ||
U30838.1 | - | 3, 4 | 237 |