Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029843.1 | NP_001025014.1 | 2 | 190 | ||
NM_001029844.1 | NP_001025015.1 | 2 | 190 | |||
NM_011705.3 | NP_035835.1 | 2 | 190 | |||
GenBank mRNA | AF080252.1 | - | 1 | 103 | ||
AF080253.1 | - | 2 | 144 | |||
AK032344.1 | - | 2 | 161 | |||
AK088230.1 | - | 2 | 120 | |||
AK088674.1 | - | 2 | 173 | |||
AK136352.1 | - | 2 | 164 | |||
AK166676.1 | - | 2 | 158 | |||
BC016676.1 | - | 2 | 225 | 3602292 | ||
BC051072.1 | - | 2 | 170 | |||
BY130767.1 | - | 2 | 190 |