Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011707.2 | NP_035837.1 | 3 | 454 | ||
GenBank mRNA | AF440693.1 | - | 3 | 224 | ||
AK011736.1 | - | 3 | 309 | |||
AK090325.1 | - | 3 | 309 | |||
AK167494.1 | - | 3 | 309 | |||
BC012690.1 | - | 3 | 300 | 4193767 | ||
CK617609.1 | - | 3 | 517 | |||
M77123.1 | - | 3 | 224 | |||
X63003.1 | - | 3 | 286 |