Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164314.1 | NP_001157786.1 | 2 | 259 | ||
NM_001164488.1 | NP_001157960.1 | 2 | 192 | |||
NM_011710.3 | NP_035840.3 | 2 | 205 | |||
GenBank mRNA | AK004541.1 | - | 2 | 193 | ||
AK150090.1 | - | 2 | 223 | |||
AK150267.1 | - | 2 | 223 | |||
AK152966.1 | - | 2 | 223 | |||
AK153337.1 | - | 2 | 231 | |||
AK183530.1 | - | 2 | 205 | |||
AK187600.1 | - | 1 | 158 | |||
AK190247.1 | - | 1 | 158 | |||
AK190313.1 | - | 1 | 158 | |||
AK190366.1 | - | 1 | 158 | |||
AK190440.1 | - | 1 | 158 | |||
AK190519.1 | - | 1 | 158 | |||
AK191285.1 | - | 1 | 158 | |||
AK191775.1 | - | 1 | 158 | |||
AK192120.1 | - | 1 | 158 | |||
AK193643.1 | - | 1 | 159 | |||
AK193869.1 | - | 1 | 158 | |||
AK194717.1 | - | 1 | 158 | |||
AK194913.1 | - | 1 | 158 | |||
AK194944.1 | - | 1 | 158 | |||
AK195085.1 | - | 1 | 158 | |||
AK195431.1 | - | 1 | 158 | |||
AK195829.1 | - | 1 | 158 | |||
AK195993.1 | - | 1 | 160 | |||
AK196253.1 | - | 1 | 158 | |||
AK196580.1 | - | 1 | 158 | |||
AK196956.1 | - | 1 | 158 | |||
AK197359.1 | - | 1 | 158 | |||
AK197539.1 | - | 1 | 158 | |||
AK197637.1 | - | 1 | 158 | |||
AK197871.1 | - | 1 | 158 | |||
AK197916.1 | - | 1 | 158 | |||
AK198390.1 | - | 1 | 156 | |||
AK198723.1 | - | 1 | 159 | |||
AK200710.1 | - | 1 | 158 | |||
AK200980.1 | - | 1 | 164 | |||
AK201133.1 | - | 1 | 159 | |||
AK202336.1 | - | 1 | 158 | |||
AK203449.1 | - | 1 | 158 | |||
AK204633.1 | - | 1 | 158 | |||
AK210160.1 | - | 1 | 158 | |||
BC003450.1 | - | 2 | 191 | 2654231 | ||
BC046232.1 | - | 2 | 198 | 6813406 | ||
BY773669.1 | - | 2 | 260 | |||
CD563585.1 | - | 2 | 179 | |||
CT010258.1 | - | 1 | 91 |