Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001652.3 | NP_001643.2 | 4 | 1622 | ||
GenBank mRNA | AL137716.1 | - | 7 | 4097 |