Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011716.2 | NP_035846.1 | 2 | 235 | ||
GenBank mRNA | AF084482.1 | - | 2 | 168 | ||
AJ011971.1 | - | 2 | 162 | |||
AK144561.1 | - | 2 | 187 | |||
AK145023.1 | - | 2 | 207 | |||
AK170180.1 | - | 2 | 190 | |||
BC046988.1 | - | 2 | 171 | 6492038 | ||
BY296480.1 | - | 2 | 218 |