Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011717.4 | NP_035847.2 | 2 | 516 | ||
NM_212438.3 | NP_997603.3 | 1 | 169 | |||
GenBank mRNA | AB012265.1 | - | 2 | 210 | ||
AB012266.1 | - | 2 | 210 | |||
AB255388.1 | - | 1 | 111 | |||
AK005208.1 | - | 2 | 288 | |||
AK133564.1 | - | 2 | 313 | |||
AK166423.1 | - | 3 | 438 | |||
BC053035.1 | - | 2 | 301 | 6400515 | ||
BU708881.1 | - | 2 | 326 | |||
BY091019.1 | - | 2 | 350 | |||
CA324749.1 | - | 2 | 284 | |||
CA325151.1 | - | 2 | 302 | |||
CA327606.1 | - | 1 | 84 | |||
CB245102.1 | - | 2 | 451 | |||
CB248991.1 | - | 2 | 516 | |||
CB249930.1 | - | 1 | 155 |