Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011718.2 | NP_035848.1 | 4 | 788 | ||
GenBank mRNA | U20658.1 | - | 4 | 603 | ||
U30464.1 | - | 4 | 786 | |||
U61970.1 | - | 3 | 521 |