Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293758.1 | NP_001280687.1 | 2, 3 | 298 | ||
NM_011722.4 | NP_035852.1 | 2, 3 | 352 | |||
GenBank mRNA | AF124788.1 | - | 2, 3 | 83 | ||
AF190796.1 | - | 2, 3 | 93 | |||
AK003618.1 | - | 2, 3 | 92 | |||
AK088312.1 | - | 2, 3 | 99 | |||
BC029249.1 | - | 2, 3 | 93 | 4486579 | ||
BY079663.1 | - | 2, 3 | 152 | |||
CA573939.1 | - | 2, 3 | 352 | |||
DV042569.1 | - | 2, 3 | 307 |