Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011731.3 | NP_035861.2 | 4, 5 | 925 | ||
GenBank mRNA | AF075261.1 | - | 4, 5 | 749 | ||
AK076806.1 | - | 5, 6 | 659 | |||
AK143893.1 | - | 4, 5 | 898 | |||
AK144038.1 | - | 4, 5 | 926 | |||
AK163340.1 | - | 5, 6 | 814 | |||
AK164706.1 | - | 4, 5 | 770 | |||
AK164713.1 | - | 4, 5 | 760 | |||
BC013484.1 | - | 4, 5 | 836 | 4242168 |