Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011752.2 | NP_035882.1 | 2, 3 | 474 | ||
GenBank mRNA | AB041612.1 | - | 2, 3 | 357 | ||
AK044472.1 | - | 2, 3 | 363 | |||
AK158995.1 | - | 2, 3 | 435 | |||
AK167849.1 | - | 2, 3 | 345 | |||
AK168061.1 | - | 2, 3 | 316 | |||
BC021397.1 | - | 2, 3 | 357 | 5067274 |