Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011772.2 | NP_035902.2 | 4 | 494 | ||
GenBank mRNA | AB017615.1 | - | 3 | 901 | ||
AK089522.1 | - | 4 | 328 | |||
AK165740.1 | - | 758 | ||||
AK170453.1 | - | 5 | 418 | |||
BC141286.1 | - | 3 | 592 | 9055898 |