Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011773.3 | NP_035903.2 | 2, 3 | 448 | ||
GenBank mRNA | AK144568.1 | - | 2, 3 | 432 | ||
AK165796.1 | - | 2, 3 | 411 | |||
BC066199.1 | - | 2, 3 | 430 | 30476359 | ||
BY718969.1 | - | 2, 3 | 451 | |||
U76007.1 | - | 2, 3 | 406 |