Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290993.1 | NP_001277922.1 | 1 | 488 | ||
NM_011774.3 | NP_035904.2 | 1 | 488 | |||
GenBank mRNA | AF003747.1 | - | 1 | 289 | ||
AK134245.1 | - | 1 | 466 | |||
AK166106.1 | - | 1 | 490 | |||
BC117997.1 | - | 1 | 346 | 40095367 |