Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289617.1 | NP_001276546.1 | 4 | 728 | ||
NM_011777.3 | NP_035907.1 | 4 | 728 | |||
GenBank mRNA | AK031415.1 | - | 4 | 526 | ||
AK035743.1 | - | 4 | 499 | |||
AK076491.1 | - | 4 | 499 | |||
AK090110.1 | - | 4 | 495 | |||
AK147812.1 | - | 4 | 729 | |||
AK170571.1 | - | 4 | 502 | |||
AK205275.1 | - | 3 | 307 | |||
X99063.1 | - | 4 | 686 | |||
Y07711.1 | - | 3 | 436 |