Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011653.2 | NP_035783.1 | 4 | 1442 | ||
GenBank mRNA | AK145404.1 | - | 4 | 1491 | ||
AK146983.1 | - | 4 | 1491 | |||
AK161783.1 | - | 2 | 1038 | |||
AK164335.1 | - | 4 | 1493 | |||
AK167687.1 | - | 4 | 1491 | |||
AK168762.1 | - | 4 | 1491 | |||
AK169610.1 | - | 4 | 1492 | |||
AK199768.1 | - | 1 | 207 | |||
AK216974.1 | - | 1 | 239 | |||
BC056169.1 | - | 4 | 1453 | 5715646 | ||
BC083343.1 | - | 4 | 1466 | 5687414 | ||
BC083344.1 | - | 4 | 1461 | 5693701 | ||
BC083345.1 | - | 4 | 1476 | 6415067 | ||
BC085256.1 | - | 4 | 1475 | 6306481 | ||
M13445.1 | - | 4 | 1442 | |||
M28729.1 | - | 1 | 635 |