Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293728.1 | NP_001280657.1 | 2 | 199 | ||
NM_011656.3 | NP_035786.1 | 3 | 274 | |||
GenBank mRNA | AF002860.1 | - | 3 | 168 | ||
AF047704.1 | - | 3 | 208 | |||
AK132488.1 | - | 3 | 206 | |||
AK171705.1 | - | 2 | 164 | |||
BC019213.1 | - | 3 | 205 | 5064658 |