Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011678.2 | NP_035808.2 | 5 | 596 | ||
GenBank mRNA | AK143582.1 | - | 5 | 531 | ||
AK149964.1 | - | 5 | 554 | |||
AK169372.1 | - | 5 | 575 | |||
AK169933.1 | - | 5 | 566 | |||
AK171271.1 | - | 5 | 566 | |||
AK171633.1 | - | 5 | 549 | |||
AK220512.1 | - | 5 | 553 | |||
BC011341.1 | - | 5 | 514 | 3661165 | ||
L00681.1 | - | 5 | 559 |