Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011680.2 | NP_035810.1 | 5, 6 | 634 | ||
GenBank mRNA | AB221562.1 | - | 5, 6 | 593 | ||
AK146871.1 | - | 5, 6 | 581 | |||
AK158906.1 | - | 3, 4 | 410 | |||
BC019729.1 | - | 1 | 14 | |||
BC082995.1 | - | 5, 6 | 634 | 6440967 | ||
U01662.1 | - | 5, 6 | 650 | |||
U01663.1 | - | 2, 3 | 187 |