Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011702.2 | NP_035832.1 | 7 | 733 | ||
GenBank mRNA | AK018599.1 | - | 7 | 734 | ||
AK043544.1 | - | 6 | 2454 | |||
BC089511.1 | - | 7 | 640 | 30249277 |