Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011716.2 | NP_035846.1 | 3, 4 | 448 | ||
GenBank mRNA | AF084482.1 | - | 3, 4 | 381 | ||
AJ011971.1 | - | 3, 4 | 375 | |||
AK144561.1 | - | 3, 4 | 400 | |||
AK145023.1 | - | 3, 4 | 420 | |||
AK170180.1 | - | 3, 4 | 403 | |||
BC046988.1 | - | 3, 4 | 384 | 6492038 | ||
CX562372.1 | - | 1, 2 | 78 |