Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002014.3 | NP_002005.1 | 1, 2 | 300 | ||
GenBank mRNA | AK313918.1 | - | 1, 2 | 265 | ||
BC001786.1 | - | 1, 2 | 255 | 3542330 | ||
BC007924.2 | - | 1, 2 | 200 | 4304462 | ||
DA392592.1 | - | 1, 2 | 300 | |||
M88279.1 | - | 1, 2 | 201 |