Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011739.3 | NP_035869.1 | 1 | 593 | ||
GenBank mRNA | AK008948.1 | - | 2 | 363 | ||
AK012540.1 | - | 2 | 328 | |||
AK145568.1 | - | 2 | 318 | |||
AK146371.1 | - | 2 | 318 | |||
AK150850.1 | - | 2 | 318 | |||
AK151437.1 | - | 2 | 367 | |||
AK151715.1 | - | 2 | 346 | |||
AK151864.1 | - | 2 | 367 | |||
AK159403.1 | - | 2 | 351 | |||
AK159817.1 | - | 2 | 367 | |||
AK163237.1 | - | 2 | 320 | |||
AK167397.1 | - | 2 | 319 | |||
AK177592.1 | - | 1 | 78 | |||
AK185231.1 | - | 1 | 80 | |||
AK190134.1 | - | 1 | 78 | |||
AK192231.1 | - | 1 | 78 | |||
AK193357.1 | - | 1 | 78 | |||
AK196448.1 | - | 1 | 78 | |||
AK198944.1 | - | 1 | 78 | |||
AK201174.1 | - | 1 | 78 | |||
BC003837.1 | - | 1 | 106 | |||
BC080802.1 | - | 2 | 308 | |||
BC085299.1 | - | 2 | 373 | |||
BC090838.1 | - | 1 | 593 | 6403848 | ||
BC106164.1 | - | 1 | 295 | 3582392 | ||
BC110680.1 | - | 1 | 301 | |||
BC132252.1 | - | 1 | 362 | 40130529 | ||
BC132530.1 | - | 1 | 362 | 40130807 | ||
D87662.1 | - | 1 | 285 | |||
U56243.1 | - | 1 | 268 | |||
U57312.1 | - | 1 | 248 |