Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011752.2 | NP_035882.1 | 3, 4 | 561 | ||
GenBank mRNA | AB041612.1 | - | 3, 4 | 444 | ||
AK044472.1 | - | 3, 4 | 450 | |||
AK158995.1 | - | 3, 4 | 522 | |||
AK167849.1 | - | 3, 4 | 432 | |||
AK168061.1 | - | 3, 4 | 403 | |||
AK218200.1 | - | 1, 2 | 36 | |||
BC021397.1 | - | 3, 4 | 444 | 5067274 |