Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011672.4 | NP_035802.3 | 10, 11 | 898 | ||
GenBank mRNA | AK010461.1 | - | 10, 11 | 867 | ||
AK012543.1 | - | 8, 9 | 842 | |||
AK088823.1 | - | 10, 11 | 867 | |||
AK147138.1 | - | 10, 11 | 881 | |||
AK169062.1 | - | 10, 11 | 880 | |||
BC006630.1 | - | 10, 11 | 855 | 3708320 | ||
U64445.1 | - | 10, 11 | 883 |