Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011674.4 | NP_035804.2 | 2 | 686 | ||
GenBank mRNA | AK078371.1 | - | 2 | 688 | ||
AK128994.1 | - | 2 | 682 | |||
AK137364.1 | - | 2 | 687 | |||
AK144005.1 | - | 2 | 686 | |||
AK163650.1 | - | 2 | 648 | |||
BC016885.1 | - | 681 | 4223057 | |||
X92122.1 | - | 1 | 1011 |