Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011701.4 | NP_035831.2 | 3 | 813 | ||
GenBank mRNA | AK012469.1 | - | 3 | 316 | ||
AK131618.1 | - | 3 | 815 | |||
AK133726.1 | - | 3 | 814 | |||
AK150270.1 | - | 3 | 509 | |||
AK151196.1 | - | 3 | 816 | |||
AK153019.1 | - | 3 | 816 | |||
AK159539.1 | - | 3 | 814 | |||
AK169181.1 | - | 3 | 815 | |||
AK170429.1 | - | 3 | 814 | |||
AK198215.1 | - | 1 | 44 | |||
AK208074.1 | - | 1 | 44 | |||
AK209588.1 | - | 1 | 44 | |||
AK209779.1 | - | 1 | 44 | |||
AK210390.1 | - | 1 | 44 | |||
AK210884.1 | - | 1 | 44 | |||
AK212337.1 | - | 1 | 44 | |||
AK212338.1 | - | 1 | 44 | |||
AK212907.1 | - | 1 | 44 | |||
AK213561.1 | - | 1 | 44 | |||
AK216501.1 | - | 1 | 44 | |||
AK217097.1 | - | 1 | 44 | |||
AK218080.1 | - | 1 | 44 | |||
AK218318.1 | - | 1 | 44 | |||
BC089335.1 | - | 3 | 809 | 6415629 | ||
M24849.1 | - | 3 | 730 | |||
M26251.1 | - | 3 | 1172 | |||
X51438.1 | - | 3 | 782 | |||
X56397.1 | - | 3 | 776 |