Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164314.1 | NP_001157786.1 | 8, 9 | 1301 | ||
NM_001164488.1 | NP_001157960.1 | 8, 9 | 1234 | |||
NM_011710.3 | NP_035840.3 | 8, 9 | 1247 | |||
GenBank mRNA | AK004541.1 | - | 8, 9 | 1235 | ||
AK007754.1 | - | 6, 7 | 863 | |||
AK150090.1 | - | 9, 10 | 1265 | |||
AK150267.1 | - | 9, 10 | 1349 | |||
AK152966.1 | - | 8, 9 | 1265 | |||
AK153337.1 | - | 8, 9 | 1272 | |||
AK187550.1 | - | 1, 2 | 156 | |||
AK189907.1 | - | 1, 2 | 156 | |||
AK190238.1 | - | 1, 2 | 156 | |||
AK196989.1 | - | 1, 2 | 156 | |||
AK198509.1 | - | 1, 2 | 156 | |||
AK198668.1 | - | 1, 2 | 157 | |||
AK202030.1 | - | 1, 2 | 156 | |||
BC003450.1 | - | 8, 9 | 1233 | 2654231 | ||
BC046232.1 | - | 8, 9 | 1240 | 6813406 | ||
CT010258.1 | - | 7, 8 | 1133 |