Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110791.1 | NP_001104261.1 | 6, 7 | 1057 | ||
NM_011750.2 | NP_035880.2 | 6, 7 | 1057 | |||
GenBank mRNA | AK133895.1 | - | 6, 7 | 846 | ||
AK146151.1 | - | 6, 7 | 818 | |||
AK147081.1 | - | 6, 7 | 826 | |||
AK152914.1 | - | 5, 6 | 759 | |||
AK154705.1 | - | 6, 7 | 825 | |||
AK157845.1 | - | 6, 7 | 826 | |||
BC009091.1 | - | 6, 7 | 802 | 3157495 | ||
BC055370.1 | - | 6, 7 | 816 | 30055109 | ||
X85802.1 | - | 6, 7 | 814 |