Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011752.2 | NP_035882.1 | 13 | 1341 | ||
GenBank mRNA | AB041612.1 | - | 13 | 1224 | ||
AK016551.1 | - | 7 | 508 | |||
AK044472.1 | - | 13 | 1230 | |||
AK158995.1 | - | 13 | 1302 | |||
AK167849.1 | - | 13 | 1212 | |||
AK168061.1 | - | 13 | 1183 | |||
AK170372.1 | - | 4 | 2004 | |||
BC021397.1 | - | 13 | 1224 | 5067274 |