Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289618.1 | NP_001276547.1 | 10 | 1977 | ||
NM_001289619.1 | NP_001276548.1 | 9 | 2160 | |||
NM_011777.3 | NP_035907.1 | 10 | 2253 | |||
GenBank mRNA | AK076491.1 | - | 10 | 2024 | ||
AK090110.1 | - | 10 | 2020 | |||
AK161709.1 | - | 10 | 1774 | |||
AK165290.1 | - | 6 | 2895 | |||
AK170571.1 | - | 10 | 2027 | |||
AK192660.1 | - | 1 | 233 | |||
AK194626.1 | - | 1 | 185 | |||
AK195064.1 | - | 1 | 273 | |||
AK197807.1 | - | 1 | 273 | |||
AK198013.1 | - | 1 | 273 | |||
AK198206.1 | - | 1 | 185 | |||
AK199933.1 | - | 1 | 189 | |||
AK200872.1 | - | 1 | 194 | |||
AK202450.1 | - | 1 | 207 | |||
AK213298.1 | - | 1 | 289 | |||
AK215885.1 | - | 1 | 273 | |||
AK217441.1 | - | 1 | 215 | |||
BC054775.1 | - | 9 | 2119 | 6808149 | ||
Y07711.1 | - | 9 | 1961 |