Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ash2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080793.2 | NP_001074262.1 | 3 | 192 | ||
NM_001286207.1 | NP_001273136.1 | 3 | 165 | |||
NM_011791.3 | NP_035921.2 | 3 | 282 | |||
GenBank mRNA | AB020983.2 | - | 3 | 264 | ||
AK087934.1 | - | 3 | 257 | |||
AK132471.1 | - | 3 | 264 | |||
AK145576.1 | - | 3 | 157 | |||
AK145790.1 | - | 3 | 169 | |||
AK145967.1 | - | 3 | 136 | |||
AK146254.1 | - | 3 | 194 | |||
AK146599.1 | - | 3 | 166 | |||
AK146756.1 | - | 3 | 157 | |||
AK146938.1 | - | 3 | 259 | |||
BC012957.1 | - | 3 | 269 | 4211433 | ||
BY266419.1 | - | 3 | 275 |