Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000139.4 | NP_000130.1 | 2 | 216 | ||
NM_001256916.1 | NP_001243845.1 | 2 | 216 | |||
XM_005273846.4 | XP_005273903.1 | 3 | 531 | |||
XM_011544850.2 | XP_011543152.1 | 3 | 149 | |||
XM_017017362.1 | XP_016872851.1 | 3 | 531 | |||
GenBank mRNA | AK304487.1 | - | 2 | 216 | ||
AK310142.1 | - | 2 | 216 | |||
BC074800.2 | - | 2 | 145 | 30915525 | ||
BC074843.2 | - | 2 | 145 | 30915222 | ||
D10583.1 | - | 2 | 164 | |||
DA972757.1 | - | 2 | 216 | |||
JF411082.1 | - | 2 | 114 |