Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282032.1 | NP_001268961.1 | 2 | 220 | ||
NM_011826.4 | NP_035956.1 | 2 | 220 | |||
GenBank mRNA | AF023482.1 | - | 2 | 192 | ||
AF023483.1 | - | 2 | 192 | |||
AF465243.1 | - | 2 | 84 | |||
AK002256.1 | - | 2 | 171 | |||
AK010633.1 | - | 2 | 204 | |||
AK088746.1 | - | 2 | 202 | |||
AK135138.1 | - | 2 | 182 | |||
BC006688.1 | - | 2 | 149 | 3582652 | ||
BC098225.1 | - | 2 | 174 | 1494110 | ||
BC147481.1 | - | 2 | 151 | 9056226 | ||
BY253818.1 | - | 2 | 209 |