Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011842.3 | NP_035972.3 | 3 | 566 | ||
GenBank mRNA | AF159259.1 | - | 3 | 441 | ||
AK077548.1 | - | 3 | 548 | |||
AK088158.1 | - | 3 | 479 | |||
AK147099.1 | - | 3 | 545 | |||
AK149834.1 | - | 3 | 612 | |||
AK157717.1 | - | 3 | 470 | |||
AK159979.1 | - | 3 | 542 | |||
BC079847.1 | - | 3 | 566 | 6419411 |