Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011859.3 | NP_035989.1 | 3 | 1114 | ||
GenBank mRNA | AF117814.1 | - | 2 | 816 | ||
BC023922.1 | - | 3 | 1070 | 5323948 |