Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289477.1 | NP_001276406.1 | 3 | 264 | ||
NM_001289478.1 | NP_001276407.1 | 3 | 451 | |||
NM_001289479.1 | NP_001276408.1 | 3 | 451 | |||
NM_011863.2 | NP_035993.1 | 2 | 274 | |||
GenBank mRNA | AK146507.1 | - | 2 | 147 | ||
AK153296.1 | - | 2 | 144 | |||
AK160875.1 | - | 3 | 355 | |||
AK168982.1 | - | 2 | 147 | |||
AK169117.1 | - | 3 | 326 | |||
AK169124.1 | - | 2 | 141 | |||
AK169204.1 | - | 3 | 326 | |||
BC066055.1 | - | 3 | 330 | 6833756 | ||
U34883.1 | - | 2 | 151 |