Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110331.1 | NP_001103801.1 | 6, 7 | 760 | ||
NM_011868.3 | NP_035998.2 | 6, 7 | 944 | |||
GenBank mRNA | AF153613.1 | - | 6, 7 | 916 | ||
AK009478.1 | - | 6, 7 | 744 | |||
AK170778.1 | - | 6, 7 | 729 | |||
BC001983.1 | - | 6, 7 | 654 | 3591615 |