Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044389.2 | NP_001037854.1 | 3 | 533 | ||
NM_011802.3 | NP_035932.2 | 3 | 533 | |||
GenBank mRNA | AF134983.1 | - | 3 | 484 | ||
AK041024.1 | - | 3 | 491 | |||
AK080339.1 | - | 3 | 458 | |||
AK087848.1 | - | 3 | 487 | |||
AK165208.1 | - | 3 | 462 | |||
BC061153.1 | - | 3 | 491 | 6593118 |