Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278075.1 | NP_001265004.1 | 3, 4 | 318 | ||
NM_011811.4 | NP_035941.2 | 3, 4 | 318 | |||
GenBank mRNA | AF123263.1 | - | 3, 4 | 318 | ||
AK010271.1 | - | 3, 4 | 281 | |||
AK012154.1 | - | 3, 4 | 281 | |||
AK045432.1 | - | 3, 4 | 270 | |||
AK076607.1 | - | 3, 4 | 280 | |||
AK134799.1 | - | 2, 3 | 224 | |||
AK165645.1 | - | 3, 4 | 185 | |||
AK167945.1 | - | 3, 4 | 281 | |||
AK168923.1 | - | 3, 4 | 262 | |||
AK169621.1 | - | 2, 3 | 225 | |||
BC002313.1 | - | 3, 4 | 177 | 3592805 | ||
BC016428.1 | - | 3, 4 | 285 | 4239906 |