Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110328.1 | NP_001103798.1 | 2 | 478 | ||
NM_001110329.1 | NP_001103799.1 | 2 | 421 | |||
NM_001110330.1 | NP_001103800.1 | 2 | 351 | |||
NM_011813.3 | NP_035943.3 | 2 | 462 | |||
GenBank mRNA | AF126746.1 | - | 2 | 412 | ||
AF126747.1 | - | 2 | 420 | |||
AK135672.1 | - | 2 | 351 | |||
AK139120.1 | - | 2 | 435 | |||
AK142205.1 | - | 2 | 371 | |||
AK162454.1 | - | 2 | 343 | |||
AK170633.1 | - | 2 | 347 | |||
AK196020.1 | - | 1 | 297 | |||
AK220212.1 | - | 2 | 351 | |||
BC006633.1 | - | 2 | 462 | 3482191 | ||
BC106113.1 | - | 2 | 337 | 6492060 | ||
BC138231.1 | - | 2 | 412 | 8861250 | ||
BC138232.1 | - | 2 | 437 | 8861251 | ||
BC145207.1 | - | 2 | 437 | 9053758 |