Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011814.2 | NP_035944.2 | 1, 2 | 436 | ||
GenBank mRNA | AK154243.1 | - | 1, 2 | 435 | ||
AK172042.1 | - | 1, 2 | 449 | |||
BC057007.1 | - | 1, 2 | 331 | 6816943 | ||
BC062971.1 | - | 1, 2 | 436 | 6415619 |