Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047537.1 | - | 4 | 514 | ||
AK089383.1 | - | 4 | 478 | |||
AK171139.1 | - | 4 | 489 | |||
AK199145.1 | - | 1 | 34 | |||
BC053416.1 | - | 3 | 338 | 6533681 | ||
CK619435.1 | - | 4 | 488 | |||
U00978.1 | - | 3 | 233 |