Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aadat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011834.2 | NP_035964.1 | 3, 4 | 466 | ||
GenBank mRNA | AF072376.1 | - | 3, 4 | 466 | ||
AK075578.1 | - | 3, 4 | 472 | |||
AK144042.1 | - | 3, 4 | 419 | |||
BC012637.1 | - | 3, 4 | 469 | 4160508 |