Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011840.2 | NP_035970.1 | 3, 4 | 840 | ||
GenBank mRNA | AB019374.1 | - | 3, 4 | 841 | ||
AK020716.1 | - | 3, 4 | 868 | |||
AK159261.1 | - | 3, 4 | 891 | |||
AK166137.1 | - | 3, 4 | 891 | |||
BC013697.1 | - | 3, 4 | 831 | |||
BC028260.1 | - | 3, 4 | 720 | 5067818 |