Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF099067.1 | - | 2 | 408 | ||
AJ223071.1 | - | 2 | 498 | |||
AK078809.1 | - | 2 | 520 | |||
AK132803.1 | - | 2 | 436 | |||
AK144130.1 | - | 2 | 382 | |||
AK147381.1 | - | 2 | 486 | |||
AK149584.1 | - | 2 | 436 | |||
AK168094.1 | - | 2 | 419 | |||
BC057939.1 | - | 2 | 410 | 5325558 | ||
Y09234.1 | - | 2 | 498 |