Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tenm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011855.4 | NP_035985.2 | 2 | 262 | ||
GenBank mRNA | AB025410.1 | - | 2 | 264 | ||
AK034286.1 | - | 4 | 1155 | |||
AK047423.1 | - | 4 | 1030 | |||
BC138860.1 | - | 1 | 245 | 8861882 | ||
BC138861.1 | - | 1 | 245 | 8861883 | ||
BY729750.1 | - | 4 | 448 |